The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Anti-ZPR1 Mouse Monoclonal Antibody
Supplier: Abgent
ABGEAT4614AEA 407 CHF InStock
ABGEAT4614A
Anti-ZPR1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ZPR1 (ZPR1 zinc finger)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty