Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-ZNF124 Mouse Monoclonal Antibody

Supplier: Abgent

ABGEAT4586AEA 407 CHF InStock
ABGEAT4586A
Anti-ZNF124 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ZNF124 (zinc finger protein 124)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty