Anti-TXNRD2 Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT4422AEA 407 CHF
ABGEAT4422A
Anti-TXNRD2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TXNRD2 (thioredoxin reductase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human