Anti-TEK Mouse Monoclonal Antibody
Lieferant: Abgent
ABGEAT4205AEA 407 CHF
ABGEAT4205A
Anti-TEK Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TEK (TEK tyrosine kinase, endothelial)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human