Anti-MAGI1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2763AEA 407 CHF
ABGEAT2763A
Anti-MAGI1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: MAGI1 (membrane associated guanylate kinase, WW and PDZ domain containing 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR