Anti-KCNMB3 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2601AEA 407 CHF
ABGEAT2601A
Anti-KCNMB3 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: KCNMB3 (potassium large conductance calcium-activated channel, subfamily M beta member 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR