Anti-KCNJ10 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2598AEA 407 CHF
ABGEAT2598A
Anti-KCNJ10 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: KCNJ10 (potassium inwardly-rectifying channel, subfamily J, member 10)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR