Anti-ITGA2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2563AEA 407 CHF
ABGEAT2563A
Anti-ITGA2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ITGA2 (integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR