Anti-ICOSLG Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2470AEA 407 CHF
ABGEAT2470A
Anti-ICOSLG Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ICOSLG (inducible T-cell co-stimulator ligand)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR