Anti-HSD3B1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2439AEA 407 CHF
ABGEAT2439A
Anti-HSD3B1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR