Anti-HRSP12 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2437AEA 407 CHF
ABGEAT2437A
Anti-HRSP12 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: HRSP12 (heat-responsive protein 12)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR