Anti-HEYL Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2360AEA 407 CHF
ABGEAT2360A
Anti-HEYL Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HEYL (hes-related family bHLH transcription factor with YRPW motif-like)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR