Anti-HERPUD1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2353AEA 407 CHF
ABGEAT2353A
Anti-HERPUD1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: HERPUD1 (homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR