Anti-HADH Mouse Monoclonal Antibody
Supplier: Abgent
ABGEAT2310AEA 407 CHF
ABGEAT2310A
Anti-HADH Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: HADH (hydroxyacyl-CoA dehydrogenase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human