Anti-GTF2H1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2287AEA 407 CHF
ABGEAT2287A
Anti-GTF2H1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GTF2H1 (general transcription factor IIH, polypeptide 1, 62kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR