Anti-GSTP1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2283AEA 407 CHF
ABGEAT2283A
Anti-GSTP1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GSTP1 (glutathione S-transferase pi 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR