Anti-GSTM5 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2281AEA 407 CHF
ABGEAT2281A
Anti-GSTM5 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GSTM5 (glutathione S-transferase mu 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR