Anti-GPR89B Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2251AEA 407 CHF
ABGEAT2251A
Anti-GPR89B Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GPR89B (G protein-coupled receptor 89B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR