Anti-GOLGA5 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2236AEA 407 CHF
ABGEAT2236A
Anti-GOLGA5 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GOLGA5 (golgin A5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR