Anti-GNAI2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2226AEA 407 CHF
ABGEAT2226A
Anti-GNAI2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GNAI2 (guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR