Anti-GIT2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2201AEA 407 CHF
ABGEAT2201A
Anti-GIT2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GIT2 (G protein-coupled receptor kinase interacting ArfGAP 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR