Anti-GAS2L3 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2156AEA 407 CHF
ABGEAT2156A
Anti-GAS2L3 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: GAS2L3 (growth arrest-specific 2 like 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR