Anti-GADD45G Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2146AEA 407 CHF
ABGEAT2146A
Anti-GADD45G Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GADD45G (growth arrest and DNA-damage-inducible, gamma)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR