Anti-FUT7 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2124AEA 407 CHF
ABGEAT2124A
Anti-FUT7 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: FUT7 (fucosyltransferase 7 (alpha (1,3) fucosyltransferase))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR