Anti-FOXC2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT2088AEA 407 CHF
ABGEAT2088A
Anti-FOXC2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: FOXC2 (forkhead box C2 (MFH-1, mesenchyme forkhead 1))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR