Anti-FKBP1B Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2056AEA 407 CHF
ABGEAT2056A
Anti-FKBP1B Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: FKBP1B (FK506 binding protein 1B, 12.6 kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR