Anti-FBXO22 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT2017AEA 407 CHF
ABGEAT2017A
Anti-FBXO22 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: FBXO22 (F-box protein 22)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR