Anti-EXOC4 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1964AEA 407 CHF
ABGEAT1964A
Anti-EXOC4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: EXOC4 (exocyst complex component 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR