Anti-ENO1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1908AEA 407 CHF
ABGEAT1908A
Anti-ENO1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ENO1 (enolase 1, (alpha))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR