Anti-ELF3 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1895AEA 407 CHF
ABGEAT1895A
Anti-ELF3 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: ELF3 (E74-like factor 3 (ets domain transcription factor, epithelial-specific ))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR