Anti-EIF2B2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1873AEA 407 CHF
ABGEAT1873A
Anti-EIF2B2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: EIF2B2 (eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR