Anti-DNASE1L1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1798AEA 407 CHF
ABGEAT1798A
Anti-DNASE1L1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: DNASE1L1 (deoxyribonuclease I-like 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR