Anti-DEGS1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1753AEA 407 CHF
ABGEAT1753A
Anti-DEGS1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DEGS1 (delta(4)-desaturase, sphingolipid 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR