Anti-DEDD Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1748AEA 407 CHF
ABGEAT1748A
Anti-DEDD Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DEDD (death effector domain containing)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR