Anti-DBP Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1712AEA 407 CHF
ABGEAT1712A
Anti-DBP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: DBP (D site of albumin promoter (albumin D-box) binding protein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR