Anti-CRYGD Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1644AEA 407 CHF
ABGEAT1644A
Anti-CRYGD Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CRYGD (crystallin, gamma D)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR