Anti-BLOC1S4 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1570AEA 407 CHF
ABGEAT1570A
Anti-BLOC1S4 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: BLOC1S4 (biogenesis of lysosomal organelles complex-1, subunit 4, cappuccino)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR