Anti-CLEC4E Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1556AEA 407 CHF
ABGEAT1556A
Anti-CLEC4E Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CLEC4E (C-type lectin domain family 4, member E)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR