Anti-CHFR Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1519AEA 407 CHF
ABGEAT1519A
Anti-CHFR Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CHFR (checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR