Anti-CHAF1A Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1512AEA 407 CHF
ABGEAT1512A
Anti-CHAF1A Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CHAF1A (chromatin assembly factor 1, subunit A (p150))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR