Anti-CCT8L2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1501AEA 407 CHF
ABGEAT1501A
Anti-CCT8L2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CCT8L2 (chaperonin containing TCP1, subunit 8 (theta)-like 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR