Anti-CDKN2D Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1483AEA 407 CHF
ABGEAT1483A
Anti-CDKN2D Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CDKN2D (cyclin-dependent kinase inhibitor 2D (p19, inhibits CDK4))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR