Anti-CDH18 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1473AEA 407 CHF
ABGEAT1473A
Anti-CDH18 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CDH18 (cadherin 18, type 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR