Anti-CDH11 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1471AEA 407 CHF
ABGEAT1471A
Anti-CDH11 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CDH11 (cadherin 11, type 2, OB-cadherin (osteoblast))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR