Anti-CD3E Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1437AEA 407 CHF
ABGEAT1437A
Anti-CD3E Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD3E (CD3e molecule, epsilon (CD3-TCR complex))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR