Anti-CD300C Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1433AEA 407 CHF
ABGEAT1433A
Anti-CD300C Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CD300C (CD300c molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR