Anti-CASP10 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT1402AEA 407 CHF
ABGEAT1402A
Anti-CASP10 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: CASP10 (caspase 10, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR