Anti-CACNA1I Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1364AEA 407 CHF
ABGEAT1364A
Anti-CACNA1I Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: CACNA1I (calcium channel, voltage-dependent, T type, alpha 1I subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR