Anti-FAM189A2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1350AEA 407 CHF
ABGEAT1350A
Anti-FAM189A2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: FAM189A2 (family with sequence similarity 189, member A2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR