Anti-C1QTNF1 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAT1338AEA 407 CHF
ABGEAT1338A
Anti-C1QTNF1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: C1QTNF1 (C1q and tumor necrosis factor related protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR